missing translation for 'onlineSavingsMsg'
Learn More

Docosahexaenoic acid, MedChemExpress

Product Code. 16452306
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
This item is not returnable. View return policy

Product Code. 16452306

Brand: Medchem Express HYB2167

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Docosahexaenoic Acid (DHA) is an omega-3 fatty acid abundantly present brain and retina. It can be obtained directly from fish oil and maternal milk.

Docosahexaenoic Acid (DHA) is an omega-3 fatty acid abundantly present brain and retina. It can be obtained directly from fish oil and maternal milk.

Specifications

Chemical Name or Material Docosahexaenoic acid
CAS 6217-54-5
Color Colorless
Purity Grade Notes Research
Molecular Formula C22H32O2
Quantity 500 mg
Synonym DHA Cervonic acid
Solubility Information DMSO : 100 mg/mL (304.42 mM; Need ultrasonic) ∣Ethanol : 50 mg/mL (152.21 mM; Need ultrasonic)
SMILES CC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CCC(O)=O
Molecular Weight (g/mol) 328.49
Formula Weight 328.49
Percent Purity 95.0%
Grade Research
Physical Form Liquid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.