missing translation for 'onlineSavingsMsg'
Learn More

Docosahexaenoic acid, MedChemExpress

Product Code. 16468856
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16468856 100 mg 100mg
16481466 200 mg 200mg
16452306 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16468856 Supplier Medchem Express Supplier No. HYB2167

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Docosahexaenoic Acid (DHA) is an omega-3 fatty acid abundantly present brain and retina. It can be obtained directly from fish oil and maternal milk.

Docosahexaenoic Acid (DHA) is an omega-3 fatty acid abundantly present brain and retina. It can be obtained directly from fish oil and maternal milk.

Tekniske data

Chemical Name or Material Docosahexaenoic acid
CAS 6217-54-5
Color Colorless
Purity Grade Notes Research
Molecular Formula C22H32O2
Quantity 100 mg
Synonym DHA Cervonic acid
Solubility Information DMSO : 100 mg/mL (304.42 mM; Need ultrasonic) ∣Ethanol : 50 mg/mL (152.21 mM; Need ultrasonic)
SMILES CC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CCC(O)=O
Molecular Weight (g/mol) 328.49
Formula Weight 328.49
Percent Purity 95.0%
Grade Research
Physical Form Liquid
For Use With (Application) Neuroscience-Neuromodulation
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.