missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOCK9 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB500-264AF405
This item is not returnable.
View return policy
Description
DOCK9 Polyclonal antibody specifically detects DOCK9 in Human samples. It is validated for Western Blot
Specifications
| DOCK9 | |
| Polyclonal | |
| Western Blot | |
| Cdc42 guanine nucleotide exchange factor zizimin-1, dedicator of cytokinesis 9, dedicator of cytokinesis protein 9, DKFZp686C11110, DKFZp686D2047, DKFZp686N04132, FLJ11949, FLJ16744, FLJ45601, KIAA1058FLJ44528, KIAA1085, ZIZ1FLJ45282, ZIZIMIN1 | |
| The immunogen recognized by this antibody maps to a region between residues 1250 and 1300 of human Dedicator of Cytokinesis 9 using the numbering given in entry NP_056111.1 (GeneID 23348). | |
| 0.1 mL | |
| Apoptosis, Cell Biology, Signal Transduction | |
| 23348 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction