Learn More
Invitrogen™ DOCK8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140262
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: INRYSSAEIR KQFTLPPNLG QYHRQSISTS GFPSLQLPQF YDPVEPVDFE GLLMTHLNSL DVQLAQELGD FTDDDLDVVF Highest antigen sequence indentity to the following orthologs: Mouse - 88%, Rat - 86%.
The Dedicator of cytokinesis protein 8 (DOCK8) is a member of the DOCK180 family of guanine nucleotide exchange factors. DOCK8 plays an essential role in humoral immune responses and is important in the proper formation of the B cell immunological synapse. Mutations in this gene result in the autosomal recessive form of the hyper-IgE syndrome.
Specifications
| DOCK8 | |
| Polyclonal | |
| Unconjugated | |
| Dock8 | |
| 1200017A24Rik; 5830472H07Rik; A130095G14Rik; AI461977; dedicator of cytokinesis 8; dedicator of cytokinesis protein 8; Dock8; epididymis luminal protein 205; HEL-205; MRD2; ZIR8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 81704 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NF50 | |
| Dock8 | |
| Recombinant protein corresponding to Human DOCK8. Recombinant protein control fragment (Product #RP-109342). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.