missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOCK7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13000-1-AP-20UL
This item is not returnable.
View return policy
Description
Multiple isoforms of DOCK 7 exist due to alternative splicing events. The DOCK 7 antibody (13000-1-AP) can recognize all isoforms.
The protein encoded by this gene is a guanine nucleotide exchange factor (GEF) that plays a role in axon formation and neuronal polarization. The encoded protein displays GEF activity toward RAC1 and RAC3 Rho small GTPases but not toward CDC42. Several transcript variants encoding different isoforms have been found for this gene.Specifications
| DOCK7 | |
| Polyclonal | |
| Unconjugated | |
| DOCK7 | |
| dedicator of cytokinesis 7, DOCK7, KIAA1771, ZIR2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 313388, 67299, 85440 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8R1A4, Q96N67 | |
| DOCK7 | |
| DOCK7 Fusion Protein Ag3671 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction