missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DOCK7 Polyclonal antibody specifically detects DOCK7 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | DOCK7 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 532 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | dedicator of cytokinesis 7, dedicator of cytokinesis protein 7, KIAA1771ZIR2 |
| Host Species | Rabbit |
| Immunogen | The immunogen for this product maps to a region between residue 175 and 225 of human Dedicator of Cytokinesis 7 using the numbering given in entry Q96N67.4 (GeneID 85440). |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?