missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DOCK5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566045
This item is not returnable.
View return policy
Description
Immunogen sequence: VELSVLFCKFI QSIPDNQLVR QKLNCMTKIV ESTLFRQSEC REVLLPLLTD QLSGQLDDNS NKPDHEASSQ LL Highest antigen sequence identity to the following orthologs - mouse 92%, rat 92%.
Guanine nucleotide exchange factor (GEF) for Rho and Rac. GEF proteins activate small GTPases by exchanging bound GDP for free GTP. Along with DOCK1, mediates CRK/CRKL regulation of epithelial and endothelial cell spreading and migration on type IV collagen (PubMed:19004829).
Specifications
| DOCK5 | |
| Polyclonal | |
| Unconjugated | |
| DOCK5 | |
| dedicator of cytokinesis 5; dedicator of cytokinesis protein 5; DOCK5; FLJ21034 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 80005 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H7D0 | |
| DOCK5 | |
| Recombinant Human DOCK5. Recombinant protein control fragment (Product #RP-106733). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction