missing translation for 'onlineSavingsMsg'
Learn More

DOCK4 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen DOCK4
Concentration 0.34 mg/mL
Applications Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16871415
View Documents
Proteintech
21861-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16861415
View Documents
Proteintech
21861-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SRC2.
Specifications

Specifications

DOCK4
Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
Rabbit
Human
Q8N1I0
9732
DOCK4 Fusion Protein Ag16516
Primary
-20°C
DOCK4
0.34 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
dedicator of cytokinesis 4, DOCK4, KIAA0716
DOCK4
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.