missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DOCK3 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA547902
This item is not returnable.
View return policy
Description
In direct ELISAs, less than 3% cross-reactivity with recombinant human (rh) DOCK1, rhDOCK2, and rhDOCK5 is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
Dedicator of cytokinesis3 (DOCK3), also named as MOCA and PBP, is a ~180 kDa protein involved in signaling transduction. It is a potential guanine nucleotide exchange factor (GEF) which activate some small GTPases by exchanging bound GDP for free GTP. DOCK3 is associated in Alzheimer disease tangles and regulates the accumulation of amyloid precursor protein and beta-amyloid. Overexpression of Dock3 in neural cells promotes axonal outgrowth downstream of brain-derived neurotrophic factor (BDNF) signaling. DOCK3 binds to and inactivates glycogen synthase kinase-3beta (GSK-3beta) at the plasma membrane, thereby promoting axon branching and microtubule assembly. By stimulating actin polymerization and microtubule assembly, DOCK3 plays important roles downstream of BDNF signaling in the CNS.
Specifications
| DOCK3 | |
| Polyclonal | |
| Unconjugated | |
| Dock3 | |
| dedicator of cytokinesis 3; dedicator of cyto-kinesis 3; dedicator of cytokinesis protein 3; DOCK3; dok-3; KIAA0299; mKIAA0299; MOCA; Modifier of cell adhesion; PBP; Presenilin-binding protein | |
| Sheep | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1795 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q8IZD9 | |
| Dock3 | |
| E. coli-derived recombinant human DOCK3 Gly418-Thr656. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction