missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DOCK3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557996
This item is not returnable.
View return policy
Description
Immunogen sequence: FEVVDSDQIS VSDLYKMHLS SRQSVQQSTS QVDTMRPRHG ETCRMPVPHH FFLSLKSFTY NTIGEDTDVF FSLYDMREGK QISERFLVRL NKNG Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
Dedicator of cytokinesis3 (DOCK3), also named as MOCA and PBP, is a ~180 kDa protein involved in signaling transduction. It is a potential guanine nucleotide exchange factor (GEF) which activate some small GTPases by exchanging bound GDP for free GTP. DOCK3 is associated in Alzheimer disease tangles and regulates the accumulation of amyloid precursor protein and beta-amyloid. Overexpression of Dock3 in neural cells promotes axonal outgrowth downstream of brain-derived neurotrophic factor (BDNF) signaling. DOCK3 binds to and inactivates glycogen synthase kinase-3beta (GSK-3beta) at the plasma membrane, thereby promoting axon branching and microtubule assembly. By stimulating actin polymerization and microtubule assembly, DOCK3 plays important roles downstream of BDNF signaling in the CNS.
Specifications
| DOCK3 | |
| Polyclonal | |
| Unconjugated | |
| Dock3 | |
| dedicator of cytokinesis 3; dedicator of cyto-kinesis 3; dedicator of cytokinesis protein 3; DOCK3; dok-3; KIAA0299; mKIAA0299; MOCA; Modifier of cell adhesion; PBP; Presenilin-binding protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1795 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IZD9 | |
| Dock3 | |
| Recombinant protein corresponding to Human DOCK3. Recombinant protein control fragment (Product #RP-96295). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction