missing translation for 'onlineSavingsMsg'
Learn More

DOCK3 Antibody [Allophycocyanin], Novus Biologicals™

Product Code. 30732573 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30732573 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732573 Supplier Novus Biologicals Supplier No. AF7134A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

DOCK3 Polyclonal antibody specifically detects DOCK3 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen DOCK3
Applications Immunohistochemistry
Classification Polyclonal
Conjugate APC
Formulation PBS
Gene Alias dedicator of cytokinesis 3, KIAA0299, modifier of cell adhesion
Host Species Sheep
Immunogen E. coli-derived recombinant human DOCK3, Gly418-Thr656, Accession # Q8IZD9
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1795
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.