Learn More
DOCK2 Mouse anti-Human, Unconjugated, Clone: 1D7A8, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66969-1-IG-150UL
 View more versions of this product
Product Code. 16839305
  • £349.00 / 150µL
Description
The DOCK2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DOCK2. This antibody recognizes human antigen. The DOCK2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
DOCK2
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, ELISA
1D7A8
Unconjugated
150 μL
1794
IgG1