Learn More
DOCK2 Mouse anti-Human, Unconjugated, Clone: 1D7A8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66969-1-IG-20UL
This item is not returnable.
View return policy
Description
The DOCK2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DOCK2. This antibody recognizes human antigen. The DOCK2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| DOCK2 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, Immunohistochemistry, ELISA | |
| 1D7A8 | |
| Unconjugated | |
| 20 μL | |
| 1794 | |
| IgG1 |