missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOCK11 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-81662AF532
This item is not returnable.
View return policy
Description
DOCK11 Polyclonal antibody specifically detects DOCK11 in Human samples. It is validated for Western Blot
Specifications
| DOCK11 | |
| Polyclonal | |
| Western Blot | |
| ACG, Activated Cdc42-associated guanine nucleotide exchange factor, Activated Cdc42-associated guanine nucleotide exchange factor, ACG, bB128O4.1, bB128O4.1.1 (novel protein), dedicator of cytokinesis 11, FLJ32122, FLJ43653, ZIZ2dedicator of cytokinesis protein 11, zizimin2, zizimin-2 | |
| The immunogen recognized by this antibody maps to a region between residue 100 and 150 of human dedicator of cytokinesis 11 using the numbering given in entry NP_653259.3 (GeneID 139818). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 139818 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction