missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DOCK1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16371095
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16371095 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16371095 Supplier Invitrogen™ Supplier No. PA584144

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: VRIMPSSLDD RRGSRPRSMV RSFTMPSSSR PLSVASVSSL SSDSTPSRPG SDGFALEPLL PKKMHSRSQD KLDKDDLEKE Highest antigen sequence indentity to the following orthologs: Rat - 94%, Mouse - 94%.

This gene encodes a loosely associated peripheral membrane protein related to the LanC family of bacterial membrane-associated proteins involved in the biosynthesis of antimicrobial peptides. This protein may play a role as a peptide-modifying enzyme component in eukaryotic cells. Previously considered a member of the G-protein-coupled receptor superfamily, this protein is now in the LanC family. Multiple alternatively spliced variants, encoding the same protein, have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DOCK1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DOCK1
Gene Accession No. Q14185
Gene Alias 180 kDa protein downstream of CRK; 9130006G06Rik; AI854900; ced5; D630004B07Rik; dedicator of cytokinesis 1; dedicator of cyto-kinesis 1; dedicator of cytokinesis protein 1; dock 180; DOCK1; DOCK180; DOwnstream of CrK; RGD1566072; RP11-82L9.1
Gene Symbols DOCK1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DOCK1. Recombinant protein control fragment (Product #RP-106449).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1793
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.