missing translation for 'onlineSavingsMsg'
Learn More

Docetaxel, MedChemExpress

Product Code. 16498056
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
Unit Size:
100mg
200mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498056 100 mg 100mg
16481356 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16498056 Supplier Medchem Express Supplier No. HYB0011
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Docetaxel (RP-56976) is a microtubule depolymerization inhibitor, with an IC50 of 0.2 μM. Docetaxel attenuates the effects of bcl-2 and bcl-xL gene expression. Docetaxel arrests the cell cycle at G2/M and leads to cell apoptosis. Docetaxel has anti-cancer activity.

Docetaxel (RP-56976) is a microtubule depolymerization inhibitor, with an IC50 of 0.2 μM. Docetaxel attenuates the effects of bcl-2 and bcl-xL gene expression. Docetaxel arrests the cell cycle at G2/M and leads to cell apoptosis. Docetaxel has anti-cancer activity.

Specifications

Chemical Name or Material Docetaxel
CAS 114977-28-5
Color White
Purity Grade Notes Research
Molecular Formula C43H53NO14
Quantity 100 mg
Synonym RP-56976
Solubility Information Ethanol : 50 mg/mL (61.89 mM; Need ultrasonic) ∣DMSO : ≥ 35 mg/mL (43.32 mM)
SMILES O=C(OC(C)(C)C)N[C@@H](C1=CC=CC=C1)[C@H](C(O[C@@H]2C(C)=C([C@@H](O)C([C@@]3(C)[C@]([C@@](CO4)(OC(C)=O)[C@@]4([H])C[C@@H]3O)([H])[C@@H]5OC(C6=CC=CC=C6)=O)=O)C(C)(C)[C@@]5(O)C2)=O)O
Molecular Weight (g/mol) 807.88
Formula Weight 807.88
Percent Purity 96.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.