missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Docetaxel, MedChemExpress
Click to view available options
Quantity:
100 mg
200 mg
Unit Size:
100mg
200mg
Description
Docetaxel (RP-56976) is a microtubule depolymerization inhibitor, with an IC50 of 0.2 μM. Docetaxel attenuates the effects of bcl-2 and bcl-xL gene expression. Docetaxel arrests the cell cycle at G2/M and leads to cell apoptosis. Docetaxel has anti-cancer activity.
Specifications
Specifications
| Chemical Name or Material | Docetaxel |
| CAS | 114977-28-5 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C43H53NO14 |
| Quantity | 200 mg |
| Synonym | RP-56976 |
| Solubility Information | Ethanol : 50 mg/mL (61.89 mM; Need ultrasonic) ∣DMSO : ≥ 35 mg/mL (43.32 mM) |
| SMILES | O=C(OC(C)(C)C)N[C@@H](C1=CC=CC=C1)[C@H](C(O[C@@H]2C(C)=C([C@@H](O)C([C@@]3(C)[C@]([C@@](CO4)(OC(C)=O)[C@@]4([H])C[C@@H]3O)([H])[C@@H]5OC(C6=CC=CC=C6)=O)=O)C(C)(C)[C@@]5(O)C2)=O)O |
| Molecular Weight (g/mol) | 807.88 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction