missing translation for 'onlineSavingsMsg'
Learn More

Docetaxel EP Impurity E, Mikromol™

Product Code. 30050708
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050708 100 mg 100mg
30050958 25 mg 25mg
30461269 4 x 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30050708 Supplier Mikromol Supplier No. MM1489.07
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Docetaxel EP Impurity E
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 10-Deacetylbaccatin-III
CAS 32981-86-5
Analyte or Component Names 10-Deacetylbaccatin-III
Shipping Condition Room Temperature
Molecular Formula C29 H36 O10
Certifications/Compliance ISO 17025
Synonym 10-O-Deacetylbaccatin III, 7,11-Methano-5H-cyclodeca[3,4]benz[1,2-b]oxet-5-one, 12b-(acetyloxy)-12-(benzoyloxy)-1,2a,3,4,4a,6,9,10,11,12,12a,12b-dodecahydro-4,6,9,11-tetrahydroxy-4a,8,13,13-tetramethyl-, (2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-, (2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-12b-(Acetyloxy)-12-(benzoyloxy)-1,2a,3,4,4a,6,9,10,11,12,12a,12b-dodecahydro-4,6,9,11-tetrahydroxy-4a,8,13,13-tetramethyl-7,11-methano-5H-cyclodeca[3,4]benz[1,2-b]oxet-5-one, 10-Deacetylbaccatin III, 10-Desacetylbaccatin III, NSC 251677, 7,11-Methano-5H-cyclodeca[3,4]benz[1,2-b]oxet-5-one, 12b-(acetyloxy)-12-(benzoyloxy)-1,2a,3,4,4a,6,9,10,11,12,12a,12b-dodecahydro-4,6,9,11-tetrahydroxy-4a,8,13,13-tetramethyl-, [2aR-(2aalpha,4beta,4abeta,6beta,9alpha,11alpha,12alpha,12aalpha,12balpha)]-, Tax-11-en-9-one, 5beta,20-epoxy-1,2alpha,4,7beta,10beta,13alpha-hexahydroxy-, 4-acetate 2-benzoate (8CI), 10beta-Deacetylbaccatin III, Docetaxel Anhydrous Impurity E, Docetaxel Impurity E
InChI Formula InChI=1S/C29H36O10/c1-14-17(31)12-29(36)24(38-25(35)16-9-7-6-8-10-16)22-27(5,23(34)21(33)20(14)26(29,3)4)18(32)11-19-28(22,13-37-19)39-15(2)30/h6-10,17-19,21-22,24,31-33,36H,11-13H2,1-5H3/t17-,18-,19+,21+,22-,24-,27+,28-,29+/m0/s1
SMILES CC(=O)O[C@@]12CO[C@@H]1C[C@H](O)[C@]3(C)[C@@H]2[C@H](OC(=O)c4ccccc4)[C@]5(O)C[C@H](O)C(=C([C@@H](O)C3=O)C5(C)C)C
Molecular Weight (g/mol) 544.59
Quantity 100 mg
Formula Weight 544.2308 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.