missing translation for 'onlineSavingsMsg'
Learn More

Docetaxel-d6 Metabolites M1 and M3 (Mixture of Diastereomers), TRC

Product Code. 30384413
Change view
Click to view available options
Quantity:
0.5 mg
5 mg
Unit Size:
0.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384413 5 mg -
30384767 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30384413 Supplier Toronto Research Chemicals Supplier No. TRCD4944075MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C43H45D6NO15
Molecular Weight (g/mol) 827.9
Synonym RPR 111026-d6
IUPAC Name (2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-12b-acetoxy-4,6,11-trihydroxy-9-(((2R,3S)-2-hydroxy-3-(4-hydroxy-5,5-bis(methyl-d3)-2-oxooxazolidin-3-yl)-3-phenylpropanoyl)oxy)-4a,8,13,13-tetramethyl-5-oxo-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-1H-7,11-methanocyclodeca[3,4]benzo[1,2-b]oxet-12-yl benzoate
SMILES O[C@H](C1=C(C)[C@@H](OC([C@H](O)[C@H](C2=CC=CC=C2)N3C(O)C(C([2H])([2H])[2H])(C([2H])([2H])[2H])OC3=O)=O)C[C@]([C@@H](OC(C4=CC=CC=C4)=O)[C@@]5([H])[C@@]6(C)[C@@H](O)C[C@]7([H])[C@@]5(OC(C)=O)CO7)(O)C1(C)C)C6=O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C43H51NO15/c1-21-25(56-36(51)31(48)29(23-14-10-8-11-15-23)44-37(52)40(5,6)59-38(44)53)19-43(54)34(57-35(50)24-16-12-9-13-17-24)32-41(7,33(49)30(47)28(21)39(43,3)4)26(46)18-27-42(32,20-55-27)58-22(2)45/h8-17,25-27,29-32,34,37,46-48,52,54H,18-20H2,1-7H3/t25-,26-,27+,29?,30+,31+,32-,34-,37?,41?,42-,43+/m0/s1/i5D3,6D3
Formula Weight 827.36
Chemical Name or Material Docetaxel-d6 Metabolites M1 and M3 (Mixture of Diastereomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.