missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Docetaxel, 98%

Product Code. p-7077609 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15316097 250 mg 250mg
15345068 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15316097 Supplier Thermo Scientific Chemicals Supplier No. 456262500
Add to basket
This item is not returnable. View return policy

CAS: 114977-28-5 | C43H53NO14 | 807.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114977-28-5
Molecular Formula C43H53NO14
Molecular Weight (g/mol) 807.88
InChI Key ZDZOTLJHXYCWBA-VCVYQWHSSA-N
Synonym docetaxel, taxotere, docetaxel anhydrous, docetaxol, emdoc, docetaxel, trihydrate, docetaxel inn, taxotere tn, unii-699121phca, txl
PubChem CID 148124
ChEBI CHEBI:4672
SMILES CC1=C2C(C(=O)C3(C(CC4C(C3C(C(C2(C)C)(CC1OC(=O)C(C(C5=CC=CC=C5)NC(=O)OC(C)(C)C)O)O)OC(=O)C6=CC=CC=C6)(CO4)OC(=O)C)O)C)O

Specifications

Melting Point 186.0°C to 192.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 250 mg
Solubility Information Solubility in water: insoluble.
Formula Weight 807.88
Percent Purity 98%
Chemical Name or Material Docetaxel
compliance-icons compliance-icons
Product Identifier
  • Docetaxel
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.