missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DOC2A Monoclonal Antibody (842213)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561881

Product Code. 30535756

  • £357.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.

There are at least two protein isoforms of the Double C2 protein, namely alpha (DOC2A) and beta (DOC2B), which contain two C2-like domains. DOC2A and DOC2B are encoded by different genes; these genes are at times confused with the unrelated DAB2 gene which was initially named DOC-2. DOC2A is mainly expressed in brain and is suggested to be involved in Ca(2+)-dependent neurotransmitter release.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DOC2A
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q14183
DOC2A
E. coli-derived recombinant human Doc2 alpha , Met1-Lys114.
100 μg
Primary
Human
Antibody
IgG2b
Immunohistochemistry (Paraffin)
842213
Unconjugated
DOC2A
Doc2; DOC2A; Doc2-alpha; double C2 domain alpha; double C2, alpha; double C2-like domain-containing protein alpha; double C2-like domains, alpha
Mouse
Protein A/G
RUO
8448
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.