missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ DOBA Medium, Powdered Media

Product Code. p-4474889 Shop All MP Biomedicals Products
Click to view available options
Quantity:
454 g
1000 g
Unit Size:
1kg
454g

Product Code. 11377799

Brand: MP Biomedicals™ 114026022

Please to purchase this item. Need a web account? Register with us today!

Can be used to create SD or SDA by combining with the appropriate amino acid mixture

Formulation

  • Use 43.7g powder liter
  • Contents/liter: DOB, 17g Agar-Y

Applications:

Yeast cell culture, Gene expression, Transcription regulation

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Molecular biology Certified™
Content And Storage 15°C to 30°C
Form Powder
Grade Molecular biology
Product Type Yeast Media
Formulation Nitrogen Sources : Ammonium Sulfate 5g/L, Carbon Source: Dextrose 20g/L, Vitamins : Biotin 0.002mg/L, Calcium Pantothenate 0.4mg/L, Folic Acid 0.002mg/L, Inositol 2mg/L, Niacin 0.4mg/L, P-Aminobenzoic Acid 0.2mg/L, Pyridoxine Hydrochloride 0.4mg/L, Riboflavin 0.2mg/L, Thiamine Hydrochloride 0.4mg/L, Trace Elements: Boric Acid 0.5mg/L, Copper Sulfate 0.04mg/L, Potassium Iodide 0.1mg/L, Ferric Chloride 0.2mg/L, Manganese Sulfate 0.4mg/L, Sodium Molybdate 0.2mg/L, Zinc Sulfate 0.4mg/L, Salts: Potassium Phosphate, Monobasic 1000mg/L, Magnesium Sulfate 500mg/L, Sodium Chloride 100mg/L, Calcium Chloride 100mg/L, Agar 17g/L
For Use With (Application) For Yeast cell culture, gene expression, transcription regulation
Quantity 454 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.