missing translation for 'onlineSavingsMsg'
Learn More

DO3A tert-Butyl ester, MedChemExpress

Product Code. 16495946
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
Unit Size:
100mg
200mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16495946 100 mg 100mg
16451046 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16495946 Supplier Medchem Express Supplier No. HY12692
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

MedChemExpress DOTA tert-Butyl ester is a benxyl derivative of the cyclic tosamide; can be nitrated directly; is more convenient to incorporate the nitro group after deprotection lithium aluminum hydride.

DOTA tert-Butyl ester is a benxyl derivative of the cyclic tosamide

Especificaciones

Chemical Name or Material DO3A tert-Butyl ester
Physical Form Solid
CAS 122555-91-3
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C26H50N4O6
Synonym DO3A tert-butyl DO3A-t-Bu-ester
Solubility Information DMSO : 62.5 mg/mL (121.43 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(OC(C)(C)C)CN1CCN(CC(OC(C)(C)C)=O)CCNCCN(CC(OC(C)(C)C)=O)CC1
Molecular Weight (g/mol) 514.7
Formula Weight 514.7
Percent Purity 98.0%
Grade Research
Mostrar más Mostrar menos

Research purposes only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.