missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNTTIP2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-84810-0.1ML
This item is not returnable.
View return policy
Description
DNTTIP2 Polyclonal specifically detects DNTTIP2 in Human samples. It is validated for Western Blot.
Specifications
| DNTTIP2 | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 30836 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| acidic 82 kDa protein mRNA, deoxynucleotidyltransferase terminal-interacting protein 2, deoxynucleotidyltransferase, terminal, interacting protein 2, ERBP, estrogen receptor binding protein, FCF2, FLJ98849, HSU15552, LPTS-interacting protein 2, LPTS-RP2, MGC163494, TdIF2, tdT-interacting factor 2 | |
| The immunogen is a synthetic peptide directed towards the middle region of human DNTTIP2 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction