missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNTTIP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA543625
This item is not returnable.
View return policy
Description
Peptide sequence: CLEQAKLLFS DGEKVIPRLT HELPGIKRGR QAEEECAHRG SPLPKKRKGR Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 100%.
RICTOR and MTOR are components of a protein complex that integrates nutrient- and growth factor-derived signals to regulate cell growth.
Specifications
| DNTTIP1 | |
| Polyclonal | |
| Unconjugated | |
| DNTTIP1 | |
| 6430706C13Rik; C20orf167; deoxynucleotidyltransferase; deoxynucleotidyltransferase terminal interacting protein 1; deoxynucleotidyltransferase terminal-interacting protein 1; deoxynucleotidyltransferase, terminal, interacting protein 1; dJ447F3.4; Dnttip1; RP3-447F3.5; Tdif1; TdT binding protein; tdT-interacting factor 1; terminal deoxynucleotidyltransferase interacting factor 1; terminal deoxynucleotidyltransferase-interacting factor 1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 116092 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9H147 | |
| DNTTIP1 | |
| Synthetic peptide directed towards the N-terminal of human DNTTIP1 (aa 121-170). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction