missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNTTIP1 Monoclonal Antibody (OTI1F4)

Mouse Monoclonal Antibody
Marca: Invitrogen™ MA525095
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
DNTTIP1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
RICTOR and MTOR are components of a protein complex that integrates nutrient- and growth factor-derived signals to regulate cell growth.
Specifica
| DNTTIP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9H147 | |
| DNTTIP1 | |
| Full length human recombinant protein of DNTTIP1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI1F4 | |
| Unconjugated | |
| DNTTIP1 | |
| 6430706C13Rik; C20orf167; deoxynucleotidyltransferase; deoxynucleotidyltransferase terminal interacting protein 1; deoxynucleotidyltransferase terminal-interacting protein 1; deoxynucleotidyltransferase, terminal, interacting protein 1; dJ447F3.4; Dnttip1; RP3-447F3.5; Tdif1; TdT binding protein; tdT-interacting factor 1; terminal deoxynucleotidyltransferase interacting factor 1; terminal deoxynucleotidyltransferase-interacting factor 1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 116092 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto