missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNTTIP1 Antibody (OTI1F4), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72352MFV500
This item is not returnable.
View return policy
Description
DNTTIP1 Monoclonal antibody specifically detects DNTTIP1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
| DNTTIP1 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry | |
| OTI1F4 | |
| Flow Cytometry | |
| C20orf167, chromosome 20 open reading frame 167, deoxynucleotidyltransferase terminal-interacting protein 1, deoxynucleotidyltransferase, terminal, interacting protein 1, dJ447F3.4, TDIF1, TdT binding protein, TdT-interacting factor 1, Terminal deoxynucleotidyltransferase-interacting factor 1 | |
| Full length human recombinant protein of human DNTTIP1 (NP_443183) produced in HEK293T cell. | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 116092 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction