missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ dNTPs Mix (10mm each)

Product Code. 11457120 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 μmol
5 x 5 μmol
Unit Size:
5µmol
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11457120 5 μmol 5µmol
12846895 5 x 5 μmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11457120 Supplier MP Biomedicals™ Supplier No. 11NTPMX100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Improves PCR efficiency, lowers denaturation temperatures and increases template lengths. MP Biomedicals dNTPs Mix (10mm each) is a ready to use solution with guaranteed purification of dNTPs (>99% pure) in order to achieve sensitive and consistent res

Manufacturing process guarantees that the dNTPs are highly purified, as contaminants can hinder PCR and result in a disappointing product yield. This product achieves sensitive and consistent results with specially prepared deoxynucleotides at a concentration of 10mm.

It is suitable for use in a range of molecular biology applications, including: PCR, real-time PCR, high fidelity and long PCR, RT-PCR, cDNA synthesis, and DNA sequencing and labeling procedures.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 mM
Content And Storage Store at -20°C.
pH 7
Quantity 5 μmol
For Use With (Application) Standard PCR, Real Time PCR (qPCR), RT-PCR
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.