missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ DNQX
GREENER_CHOICE

Product Code. 30001058
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001058 10 mg 10mg
30001059 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001058 Supplier Tocris Bioscience™ Supplier No. 0189/10
  • ONLINE EXCLUSIVE PRICE
    £86.00
    £80.85 / 10mg
    Save £5.15
    5% Off
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Selective non-NMDA iGluR antagonist

Selective non-NMDA receptor antagonist.

Water-soluble Salt also available.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2379-57-9
Molecular Formula C8H4N4O6

Specifications

Quantity 10 mg
Formula Weight 252.14
Purity >98%
Molecular Weight (g/mol) 252.14
Chemical Name or Material DNQX
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.