missing translation for 'onlineSavingsMsg'
Learn More

DNQX, MedChemExpress

Product Code. 16496626
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
200mg
25mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496626 100 mg 100mg
16491156 200 mg 200mg
16403466 25 mg 25mg
16402427 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16496626 Supplier Medchem Express Supplier No. HY15067
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress DNQX (FG 9041), a quinoxaline derivative, is a selective, potent competitive non-NMDA glutamate receptor antagonist (IC50s = 0.5, 2 and 40 μM for AMPA, kainate and NMDA receptors, respectively).

DNQX (FG 9041), a quinoxaline derivative, is a selective, potent competitive non-NMDA glutamate receptor antagonist (IC50s = 0.5, 2 and 40 μM for AMPA, kainate and NMDA receptors, respectively).

Specifications

Chemical Name or Material DNQX
Physical Form Powder
CAS 2379-57-9
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C8H4N4O6
For Use With (Application) Cancer-programmed cell death
Synonym FG 9041
Solubility Information DMSO : ≥ 35 mg/mL (138.81 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C1C(NC2=CC([N+]([O-])=O)=C([N+]([O-])=O)C=C2N1)=O
Molecular Weight (g/mol) 252.14
Formula Weight 252.14
Percent Purity 98.01%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.