Learn More
Invitrogen™ DNMT3B Recombinant Rabbit Monoclonal Antibody (7Y9S2)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA551459
Description
Immunogen sequence: MKGDTRHLNG EEDAGGREDS ILVNGACSDQ SSDSPPILEA IRTPEIRGRR SSSRLSKREV SSLLSYTQDL TGDGDGEDGD GSDTPVMPKL FRETRTRSES PAVRTRNNNS VSSRERHRPS PRSTRGRQGR NHVDESPVEF PATRSLRRRA TASAGTPWPS PPSSYLTIDL TDDTEDTHGT PQSSSTPYAR LAQDSQQGGM ESPQVEADSG DGDSSEYQDG KEFGIGDLVW GKIKGFSWWP AMVVSWKATS KRQAMSGMRW VQWFGDGKFS EVSADKLVAL GLFSQHFNLA TFNKLVSYRK AMYHALEKAR VRAGKTFPSS PGDSLEDQLK PMLEWAHGGF KPTGIEGLKP NNTQPVVNKS KVRRAGSRKL ESRKYENKTR RRTADDSATS DYCPAPKRLK TNCYNNGKDR.
CpG methylation is an epigenetic modification that is important for embryonic development, imprinting, and X-chromosome inactivation. Studies in mice have demonstrated that DNA methylation is required for mammalian development. This gene encodes a DNA methyltransferase which is thought to function in de novo methylation, rather than maintenance methylation. The protein localizes primarily to the nucleus and its expression is developmentally regulated. Mutations in this gene cause the immunodeficiency-centromeric instability-facial anomalies (ICF) syndrome. Six alternatively spliced transcript variants have been described. The full length sequences of variants 4 and 5 have not been determined.
Specifications
| DNMT3B | |
| Recombinant Monoclonal | |
| 0.21 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| O88509, Q9UBC3 | |
| Dnmt3b | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1- 410 of human DNMT3B (NP_612564.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| Liquid |
| ChIP Assay, ChIP sequencing (ChIP-seq), ELISA, Western Blot | |
| 7Y9S2 | |
| Unconjugated | |
| Dnmt3b | |
| DNA (cytosine-5-)-methyltransferase 3 beta; DNA (cytosine-5)-methyltransferase 3B; DNA methyltransferase 3 beta; DNA methyltransferase 3B; DNA methyltransferase HsaIIIB; DNA methyltransferase MmuIIIB; DNA MTase HsaIIIB; DNA MTase MmuIIIB; Dnmt3; DNMT3B; ICF; ICF1; M.HsaIIIB; m.MmuIIIB; MGC1090; MmuIIIB | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 13436, 1789 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.