missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DNMT3B Recombinant Rabbit Monoclonal Antibody (1I5F7)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA558044

 View more versions of this product

Product Code. 30282705

  • £363.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MKGDTRHLNG EEDAGGREDS ILVNGACSDQ SSDSPPILEA IRTPEIRGRR SSSRLSKREV SSLLSYTQDL TGDGDGEDGD GSDTPVMPKL FRETRTRSES PAVRTRNNNS VSSRERHRPS PRSTRGRQGR NHVDESPVEF PATRSLRRRA TASAGTPWPS PPSSYLTIDL TDDTEDTHGT PQSSSTPYAR LAQDSQQGGM ESPQVEADSG DGDSSEYQDG KEFGIGDLVW GKIKGFSWWP AMVVSWKATS KRQAMSGMRW VQWFGDGKFS EVSADKLVAL GLFSQHFNLA TFNKLVSYRK AMYHALEKAR VRAGKTFPSS PGDSLEDQLK PMLEWAHGGF KPTGIEGLKP NNTQPVVNKS KVRRAGSRKL ESRKYENKTR RRTADDSATS DYCPAPKRLK TNCYNNGKDR.

CpG methylation is an epigenetic modification that is important for embryonic development, imprinting, and X-chromosome inactivation. Studies in mice have demonstrated that DNA methylation is required for mammalian development. This gene encodes a DNA methyltransferase which is thought to function in de novo methylation, rather than maintenance methylation. The protein localizes primarily to the nucleus and its expression is developmentally regulated. Mutations in this gene cause the immunodeficiency-centromeric instability-facial anomalies (ICF) syndrome. Six alternatively spliced transcript variants have been described. The full length sequences of variants 4 and 5 have not been determined.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DNMT3B
Recombinant Monoclonal
1.06 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
O88509
Dnmt3b
Recombinant fusion protein containing a sequence corresponding to amino acids 1-410 of human DNMT3B (NP_008823.1).
100 μL
Primary
Mouse
Antibody
IgG
ELISA, Western Blot
1I5F7
Unconjugated
Dnmt3b
DNA (cytosine-5-)-methyltransferase 3 beta; DNA (cytosine-5)-methyltransferase 3B; DNA methyltransferase 3 beta; DNA methyltransferase 3B; DNA methyltransferase HsaIIIB; DNA methyltransferase MmuIIIB; DNA MTase HsaIIIB; DNA MTase MmuIIIB; Dnmt3; DNMT3B; ICF; ICF1; M.HsaIIIB; m.MmuIIIB; MGC1090; MmuIIIB
Rabbit
Protein A
RUO
13436
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.