missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNMT3B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585549
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Predicted reactivity: Pig (85%). Positive Control: HeLa, HeLa nuclear, GFP-tagged DNMT3B-transfested 293T. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CpG methylation is an epigenetic modification that is important for embryonic development, imprinting, and X-chromosome inactivation. Studies in mice have demonstrated that DNA methylation is required for mammalian development. This gene encodes a DNA methyltransferase which is thought to function in de novo methylation, rather than maintenance methylation. The protein localizes primarily to the nucleus and its expression is developmentally regulated. Mutations in this gene cause the immunodeficiency-centromeric instability-facial anomalies (ICF) syndrome. Six alternatively spliced transcript variants have been described. The full length sequences of variants 4 and 5 have not been determined.
Specifications
| DNMT3B | |
| Polyclonal | |
| Unconjugated | |
| Dnmt3b | |
| DNA (cytosine-5-)-methyltransferase 3 beta; DNA (cytosine-5)-methyltransferase 3B; DNA methyltransferase 3 beta; DNA methyltransferase 3B; DNA methyltransferase HsaIIIB; DNA methyltransferase MmuIIIB; DNA MTase HsaIIIB; DNA MTase MmuIIIB; Dnmt3; DNMT3B; ICF; ICF1; M.HsaIIIB; m.MmuIIIB; MGC1090; MmuIIIB | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1789, 444985 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1.29 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q9UBC3 | |
| Dnmt3b | |
| Recombinant protein encompassing a sequence within the N-terminus region of human DNMT3B. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction