missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DNMT3A Recombinant Rabbit Monoclonal Antibody (K01_2M16)

Product Code. 30828083
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30828083 50 μL
30827162 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30828083 Supplier Invitrogen™ Supplier No. MA565507

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

DNMT3A Recombinant Monoclonal Antibody for Western Blot

Methylation of DNA at cytosine residues plays an important role in regulation of gene expression, genomic imprinting and is essential for mammalian development. Hypermethylation of CpG islands in tumor suppressor genes or hypomethylation of bulk genomic DNA may be linked with development of cancer. To date, 3 families of mammalian DNA methyltransferase genes have been identified which include Dnmt1, Dnmt2 and Dnmt3. Dnmt1 is constitutively expressed in proliferating cells and inactivation of this gene causes global demethylation of genomic DNA and embryonic lethality. Dnmt2 is expressed at low levels in adult tissues and its inactivation does not affect DNA methylation or maintenance of methylation. The Dnmt3 family members, Dnmt3a and Dnmt3b, are strongly expressed in ES cells but their expression is down regulated in differentiating ES cells and is low in adult somatic tissue. Recently, it has been shown that naturally occurring mutations of Dnmt3b gene occurs in patients with a rare autosomal recessive disorder, termed ICF (immunodeficiency, centromeric instability, and facial anomalies) syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNMT3A
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_2M16
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene DNMT3A
Gene Accession No. Q9Y6K1
Gene Alias AU015806; DNA (cytosine-5-)-methyltransferase 3 alpha; DNA (cytosine-5)-methyltransferase 3A; DNA cytosine methyltransferase 3A2; DNA methyltransferase 3 alpha; DNA methyltransferase 3A; DNA methyltransferase HsaIIIA; DNA methyltransferase MmuIIIA; DNA MTase HsaIIIA; DNA MTase MmuIIIA; Dnmt3a; DNMT3A2; M.HsaIIIA; MmuIIIA; TBRS; W91664
Gene Symbols DNMT3A
Host Species Rabbit
Immunogen A synthetic peptide of human Dnmt3a.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1788
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.