missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNMT3A Recombinant Rabbit Monoclonal Antibody (23GB2230), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | DNMT3A |
|---|---|
| Clone | 23GB2230 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088022
![]() |
Invitrogen™
MA552484 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088378
|
Invitrogen™
MA552485 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DNMT3A Recombinant Monoclonal antibody specifically detects DNMT3A in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| DNMT3A | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O88508, Q1LZ53, Q9Y6K1 | |
| 13435, 1788, 444984 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB2230 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AU015806; DNA (cytosine-5-)-methyltransferase 3 alpha; DNA (cytosine-5)-methyltransferase 3A; DNA cytosine methyltransferase 3A2; DNA methyltransferase 3 alpha; DNA methyltransferase 3A; DNA methyltransferase HsaIIIA; DNA methyltransferase MmuIIIA; DNA MTase HsaIIIA; DNA MTase MmuIIIA; Dnmt3a; DNMT3A2; M.HsaIIIA; MmuIIIA; TBRS; W91664 | |
| A synthesized peptide derived from human Dnmt3a (609-624AA). | |
| Primary | |
| -20°C | |
| DNMT3A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title