missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNMT3A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19366-1-AP-20UL
This item is not returnable.
View return policy
Description
TREF1 encodes a zinc-finger transcriptional regulating protein which interacts with CBP/p300 to regulate the human gene CYP11A1.Specifications
| DNMT3A | |
| Polyclonal | |
| Unconjugated | |
| DNMT3A | |
| DNA methyltransferase HsaIIIA, DNA MTase HsaIIIA, DNMT3A, DNMT3A2, M.HsaIIIA | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13435, 1788, 444984 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O88508, Q1LZ53, Q9Y6K1 | |
| DNMT3A | |
| DNMT3A Fusion Protein Ag7015 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction