missing translation for 'onlineSavingsMsg'
Learn More

DNMT3A Rabbit anti-Human, Mouse, Rat, Clone: 7L9O3, Novus Biologicals™

Product Code. 18369984 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18369984 100 μg 100µL
18370107 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18369984 Supplier Novus Biologicals Supplier No. NBP315844100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

DNMT3A Monoclonal antibody specifically detects DNMT3A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNMT3A
Applications Western Blot, Immunoprecipitation, KnockDown
Classification Monoclonal
Clone 7L9O3
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias DNA (cytosine-5-)-methyltransferase 3 alpha, DNA (cytosine-5)-methyltransferase 3A, DNA cytosine methyltransferase 3A2, DNA Methyltransferase 3 Alpha, DNA methyltransferase HsaIIIA, DNA MTase HsaIIIA, Dnmt3a, DNMT3A2, EC 2.1.1.37, M.HsaIIIA, TBRS
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 550-650 of human DNMT3A (Q9Y6K1).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, Chromatin Research, Epigenetics, Innate Immunity, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 1788
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.