missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNMT3A Antibody (64B1446), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NB120-13888UV
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
DNMT3A Monoclonal specifically detects DNMT3A in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Whole-Mount.
Specifica
| DNMT3A | |
| Monoclonal | |
| DyLight 350 | |
| Q9Y6K1 | |
| DNMT3A | |
| This DNMT3A antibody was raised against bacteria expressed recombinant mouse Dnmt3a. The epitope was found to lie near the C-terminus (a.a. 705-908), see Chen et (2002) for details. | |
| 0.1 mL | |
| Chromatin Modifiers, Chromatin Research, DNA Methyltransferases, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics | |
| 1788 | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 64B1446 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Whole-Mount | |
| DNA (cytosine-5-)-methyltransferase 3 alpha, DNA (cytosine-5)-methyltransferase 3A, DNA cytosine methyltransferase 3A2, DNA methyltransferase HsaIIIA, DNA MTase HsaIIIA, Dnmt3a, DNMT3A2, EC 2.1.1.37, M.HsaIIIA | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Cross-reacts with Human (weakly) and Mouse. Not yet tested in other species. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto