missing translation for 'onlineSavingsMsg'
Learn More

Dnmt2 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Product Code. 18365505 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18365505 25 μg 25µL
18345095 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18365505 Supplier Novus Biologicals Supplier No. NBP31866325UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Dnmt2 Polyclonal antibody specifically detects Dnmt2 in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dnmt2
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:100-1:1000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200, Immunohistochemistry-Frozen
Formulation Whole antiserum
Gene Accession No. O14717
Gene Alias DMNT2, DNA (cytosine-5-)-methyltransferase 2, DNA (cytosine-5)-methyltransferase-like protein 2, DNA methyltransferase homolog HsaIIP, DNA methyltransferase-2, DNA MTase homolog HsaIIP, Dnmt2, EC 2.1.1, EC 2.1.1.29, M.HsaIIP, MHSAIIP, PuMet, RNMT1, tRNA (cytosine-5-)-methyltransferase, tRNA aspartic acid methyltransferase 1
Host Species Rabbit
Immunogen Polyclonal antibody raised in rabbit against mouse Dnmt2 (DNA methyltransferase 2), using a KLHconjugated synthetic peptide containing an amino acid sequence from the N-terminal part of the protein (1).
Purification Method Whole Antiserum
Quantity 25 μg
Regulatory Status RUO
Research Discipline Chromatin Research, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 1787
Target Species Mouse
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Antisera
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.