missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNMT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19221-1-AP-150UL
This item is not returnable.
View return policy
Description
DNMT2 encodes a member of the solute carrier family 11 protein family. The product of this gene transports divalent metals and is involved in iron absorption. Mutations in this gene are associated with hypochromic microcytic anemia with iron overload. A related solute carrier family 11 protein gene is located on chromosome 2. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
| DNMT2 | |
| Polyclonal | |
| Unconjugated | |
| TRDMT1 | |
| DNA MTase homolog HsaIIP, DNMT2, M.HsaIIP, PuMet, RNMT1, TRDMT1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13434, 1787, 291324 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O14717, O55055, Q4G073 | |
| Trdmt1 | |
| DNMT2 Fusion Protein Ag5571 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction