missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dnmt2 Antibody (102B1259.2), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB200-587R
This item is not returnable.
View return policy
Description
Dnmt2 Monoclonal specifically detects Dnmt2 in Human, Mouse, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| Dnmt2 | |
| Monoclonal | |
| DyLight 550 | |
| Q6ICS7 | |
| TRDMT1 | |
| This antibody was raised against synthetic peptides corresponding to mouse Dnmt2. | |
| 0.1 mL | |
| Chromatin Modifiers, Chromatin Research, Epigenetics | |
| 1787 | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| 102B1259.2 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| DMNT2, DNA (cytosine-5-)-methyltransferase 2, DNA (cytosine-5)-methyltransferase-like protein 2, DNA methyltransferase homolog HsaIIP, DNA methyltransferase-2, DNA MTase homolog HsaIIP, Dnmt2, EC 2.1.1, EC 2.1.1.29, M.HsaIIP, MHSAIIP, PuMet, RNMT1, tRNA (cytosine-5-)-methyltransferase, tRNA aspartic acid methyltransferase 1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| In mouse Dnmt2 transfected cell lysate, a 42 kDa band is oberseved. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction