missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNMT1 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24206-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a subunit of the augmin complex, which regulates centrosome and mitotic spindle integrity, and is necessary for the completion of cytokinesis. The encoded protein was identified by interaction with ubiquitin C-terminal hydrolase 37. Alternative splicing results in multiple transcript variants.Specifications
| DNMT1 | |
| Polyclonal | |
| Unconjugated | |
| DNMT1 | |
| AIM, CXXC9, DNA MTase HsaI, DNMT, DNMT1, MCMT | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1786, 84350 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.41 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P26358, Q9Z330 | |
| DNMT1 | |
| DNMT1 Fusion Protein Ag19116 | |
| 20 μL | |
| Primary | |
| Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction