missing translation for 'onlineSavingsMsg'
Learn More

DNMT1 Rabbit anti-Human, Mouse, Rat, Clone: 8E3H1, Novus Biologicals™

Product Code. 18322085
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18322085 20 μg 20µL
18396963 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18322085 Supplier Novus Biologicals Supplier No. NBP31585620UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

DNMT1 Monoclonal antibody specifically detects DNMT1 in Human, Mouse, Rat samples. It is validated for Western Blot, ChIP assay, Immunofluorescence, Immunoprecipitation, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNMT1
Applications ChIP Assay, Immunofluorescence, Immunoprecipitation, KnockDown
Classification Monoclonal
Clone 8E3H1
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:1000, Chromatin Immunoprecipitation 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:500 - 1:1000, Knockout Validated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias AIM, CXXC9CXXC finger protein 9, CXXC-type zinc finger protein 9, DNA (cytosine-5-)-methyltransferase 1, DNA (cytosine-5)-methyltransferase 1, DNA methyltransferase 1, DNA methyltransferase HsaI, DNA MTase HsaI, DNMT, Dnmt1, EC 2.1.1.37, FLJ16293, m.HsaI, MCMTMGC104992
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human DNMT1 (NP_001370.1). MPARTAPARVPTLAVPAISLPDDVRRRLKDLERDSLTEKECVKEKLNLLHEFLQTEIKNQLCDLETKLRKEELSEEGYLAKVKSLLNKDLSLENGAHAYN
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Chromatin Research, Core ESC Like Genes, Epigenetics, Stem Cell Markers, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 1786
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.