missing translation for 'onlineSavingsMsg'
Learn More

DNMT1 Antibody (4G11-C7), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-59344-25ug

 View more versions of this product

Product Code. 18666677

  • £177.00 / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

DNMT1 Monoclonal specifically detects DNMT1 in Human, Mouse, Fish, Zebrafish samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DNMT1
Monoclonal
Unconjugated
PBS (pH 7.4), 50% glycerol with 0.09% Sodium Azide
DNMT1
Raised against a partial protein corresponding to amino acids 620-950 of human DNMT1
25 μg
Angiogenesis, Cancer, Chromatin Modifiers, Chromatin Research, Core ESC Like Genes, DNA Methyltransferases, Epigenetics, Phospho Specific, Stem Cell Markers, Transcription Factors and Regulators
1786
Human, Mouse, Fish, Zebrafish
Purified
Western Blot, Immunohistochemistry, Immunoprecipitation, ChIP Assay
4G11-C7
Western Blot 1:1000, Immunohistochemistry 1:1000, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP) 1:10-1:500
AIM, CXXC9CXXC finger protein 9, CXXC-type zinc finger protein 9, DNA (cytosine-5-)-methyltransferase 1, DNA (cytosine-5)-methyltransferase 1, DNA methyltransferase 1, DNA methyltransferase HsaI, DNA MTase HsaI, DNMT, Dnmt1, EC 2.1.1.37, FLJ16293, m.HsaI, MCMTMGC104992
Mouse
Protein G purified
RUO
Primary
Detects 180kDa. It will cross-react with mouse DNMT1.
Store at -20°C.
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only