missing translation for 'onlineSavingsMsg'
Learn More

DNMT1 Antibody (11H8), Novus Biologicals™

Mouse Monoclonal Antibody

£171.00 - £424.00

Specifications

Antigen DNMT1
Clone 11H8
Concentration 1 mg/mL
Dilution Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100
Applications Western Blot, Immunocytochemistry, Immunofluorescence
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18214044
View Documents
Novus Biologicals
NBP2-59694
100 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
18655408
View Documents
Novus Biologicals
NBP2-59694-25ug
25 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

DNMT1 Monoclonal specifically detects DNMT1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

DNMT1
1 mg/mL
Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
Mouse
Angiogenesis, Cancer, Chromatin Modifiers, Chromatin Research, Core ESC Like Genes, DNA Methyltransferases, Epigenetics, Phospho Specific, Stem Cell Markers, Transcription Factors and Regulators
PBS, 0.05% BSA, 50% glycerol with 0.05% Sodium Azide
1786
Raised against a synthetic peptide corresponding to amino acids 637-650 of human DNMT1
Primary
It will cross-react with mouse DNMT1.
11H8
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100
Monoclonal
Purified
RUO
Human, Mouse
AIM, CXXC9CXXC finger protein 9, CXXC-type zinc finger protein 9, DNA (cytosine-5-)-methyltransferase 1, DNA (cytosine-5)-methyltransferase 1, DNA methyltransferase 1, DNA methyltransferase HsaI, DNA MTase HsaI, DNMT, Dnmt1, EC 2.1.1.37, FLJ16293, m.HsaI, MCMTMGC104992
DNMT1
IgG1
Protein G purified
180 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only (RUO)

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.