missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DNMBP Polyclonal antibody specifically detects DNMBP in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | DNMBP |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 650 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | dynamin binding protein, dynamin-binding protein, KIAA1010ARHGEF36, Scaffold protein Tuba, TUBA |
| Host Species | Rabbit |
| Immunogen | The immunogen for this product maps to a region between residue 75 and 125 of human Scaffold Protein TUBA using the numbering given in entry NP_056036.1 (GeneID 23268). |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?