missing translation for 'onlineSavingsMsg'
Learn More

DNM2 Mouse, anti-Human, CoraLite Plus 647, Clone: 1B8H3, Proteintech

Product Code. 18050736 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18050736 20 μL 20µL
18047405 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18050736 Supplier Proteintech Supplier No. CL6476820920UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The DNM2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DNM2. This antibody recognizes human, mouse, rat, pig, rabbit antigen. The DNM2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 647.

Specifications

Antigen DNM2
Applications Immunofluorescence
Clone 1B8H3
Concentration 1000 μg/mL
Conjugate CoraLite Plus 647
Gene Symbols CMTDI1, CMTDIB, DI CMTB, DNM2, DYN2, dynamin 2, DYNII
Host Species Mouse
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 1785
Target Species Human, Mouse, Rat, Pig, Rabbit
Content And Storage Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Antibody Molecular Weight 155 kDa
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.