missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DNHD1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA584355

Product Code. 16381305

  • £478.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: CWHSLFKIQN RLAAMEDTST QGCQPVEITH LYPSGLSPQE FLGWLEGSCW HHGIFPKVLR AAGQCNNMGQ KRQTEESIGI QHWIICDGAS NGAWLD Highest antigen sequence indentity to the following orthologs: Mouse - 65%, Rat - 65%.

DNHD1 is a protein coding gene. Gene ontology (GO) annotation include dynein complex; extracellular exosome.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DNHD1
Polyclonal
Unconjugated
DNHD1
8030491N06Rik; C11orf47; CCDC35; coiled-coil domain containing 35; DHCD1; DKFZp686J0796; DNHD1; DNHD1 variant protein; DNHD1L; dynein heavy chain domain 1; dynein heavy chain domain 1-like protein; Dynein heavy chain domain-containing protein 1; FLJ32752; FLJ35709; FLJ46184; Protein CCDC35; UNQ5781/PRO12970
Rabbit
Antigen affinity chromatography
RUO
144132
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunofluorescence
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q96M86
DNHD1
Recombinant protein corresponding to Human DNHD1. Recombinant protein control fragment (Product #RP-106792).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.