missing translation for 'onlineSavingsMsg'
Learn More

DNER Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£328.00 - £443.00

Specifications

Antigen DNER
Concentration 0.05mg/mL
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:10 - 1:20
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18411561
View Documents
Novus Biologicals
NBP1-88665-25ul
25 μL
£328.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18216218
View Documents
Novus Biologicals
NBP1-88665
0.1 mL
£443.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

DNER Polyclonal specifically detects DNER in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

DNER
Immunohistochemistry, Immunohistochemistry-Paraffin 1:10 - 1:20
Polyclonal
Rabbit
Growth and Development, Neuronal Cell Markers
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
92737
This antibody was developed against Recombinant Protein corresponding to amino acids:GNASSNSSAGGRLVSFEVPQNTSVKIRQDATASLILLWKVTATGFQQCSLIDGRSVTPLQASGGLVLLEEMLALGNNHFIGFVNDSVTKSIVALRLTLVVKVSTCVPGESHANDLECSGKGKCTTKP
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
0.05mg/mL
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Unconjugated
RUO
Human
bet, delta and Notch-like epidermal growth factor-related receptor, delta/notch-like EGF repeat containing, delta-notch-like EGF repeat-containing transmembrane, H_NH0150O02.1, UNQ26, WUGSC:H_NH0150O02.1
DNER
IgG
Affinity Purified
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.