missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dnd1 Antibody (3H5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00373863-M01
This item is not returnable.
View return policy
Description
Dnd1 Monoclonal antibody specifically detects Dnd1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| Dnd1 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| dead end homolog 1 (zebrafish), dead end protein homolog 1, RBMS4MGC34750, RNA-binding motif, single-stranded-interacting protein 4 | |
| DND1 (NP_919225, 167 a.a. ∽ 260 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. EARLLPSPGPAPGQIALLKFSSHRAAAMAKKALVEGQSHLCGEQVAVEWLKPDLKQRLRQQLVGPFLRSPQPEGSQLALARDKLGFQGARATLQ | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| 3H5 | |
| Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence | |
| NP_919225 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 373863 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction